Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF786 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554435
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554435 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554435 Supplier Invitrogen™ Supplier No. PA554435
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EGIPGPRNLD LPGLWDVPAW ESTQHPWPVC GESCWENNHL VMHQRGHSKD RTRRAWEKFN KRAETQMPWS SPRVQRH Highest antigen sequence identity to the following orthologs: Mouse - 45%, Rat - 40%.

Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Krüppeltype DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. ZNF786 (zinc finger protein 786) is a 782 amino acid protein that belongs to the Krüppel C2H2-type zinc-finger protein family and is thought to function in transcriptional regulation. Localizing to nucleus, ZNF786 contains 16 C2H2-type zinc fingers, a single KRAB domain and is encoded by a gene that maps to human chromosome 7q36.1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF786
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF786
Gene Accession No. Q8N393
Gene Alias A730012O14Rik; Zfp786; Zinc finger protein 786; Znf786
Gene Symbols ZNF786
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF786. Recombinant protein control fragment (Product #RP-92846).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 136051
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.