Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF787 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557174
Description
Immunogen sequence: MELREEAWSP GPLDSEDQQM ASHENPVDIL IMDDDDVPSW PPTKLS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 98%.
May be involved in transcriptional regulation.
Specifications
ZNF787 | |
Polyclonal | |
Unconjugated | |
ZNF787 | |
2210018M03Rik; TIP20; transcription termination factor I interacting peptide 20; TTF-I-interacting peptide 20; Zfp787; Zinc finger protein 787; ZNF787 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
126208 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6DD87 | |
ZNF787 | |
Recombinant protein corresponding to Human ZNF787. Recombinant protein control fragment (Product #RP-97578). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction