Learn More
ZNF787 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303281AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
May be involved in transcriptional regulation.Specifications
ZNF787 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q8BIF9, Q6DD87 | |
Zfp787, ZNF787 | |
Between 1 and 50 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ZNF787 | |
Zinc finger protein 787, TTF-I-interacting peptide 20, ZNF787 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
126208, 67109 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.