Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF79 Antibody (OTI3D2), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274955C
Description
ZNF79 Monoclonal specifically detects ZNF79 in Human samples. It is validated for Western Blot.Specifications
ZNF79 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ZNF79 | |
Full length recombinant protein of human ZNF79 (NP_009066) produced in E.coli. | |
0.1 mL | |
7633.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI3D2 | |
Western Blot | |
pT7, zinc finger protein 79, zinc finger protein 79 (pT7), ZNFpT7 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction