Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF790 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569742
Description
This target displays homology in the following species: Human: 100%.
ZNF790 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.
Specifications
ZNF790 | |
Polyclonal | |
Unconjugated | |
ZNF790 | |
FLJ20350; MGC62100; zinc finger protein 790; ZNF790 | |
Rabbit | |
Affinity chromatography | |
RUO | |
388536 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6PG37 | |
ZNF790 | |
synthetic peptide directed towards the N terminal region of human ZNF790. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction