Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF790 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584449
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584449 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584449 Supplier Invitrogen™ Supplier No. PA584449
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CKKSSHIFSH HSYFTEQKIH NSANLCEWTD YGNTFSHESN FAQHQNIYTF EKSYEFKDFE KA Highest antigen sequence indentity to the following orthologs: Rat - 44%, Mouse - 44%.

ZNF790 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF790
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF790
Gene Accession No. Q6PG37
Gene Alias FLJ20350; MGC62100; zinc finger protein 790; ZNF790
Gene Symbols ZNF790
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF790. Recombinant protein control fragment (Product #RP-101696).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388536
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.