Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                ZNF791 Antibody, Novus Biologicals™
                                
                                
                                
                                
                            
                            
                            
                                
                                    
Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | ZNF791 | 
|---|---|
| Applications | Western Blot | 
| Classification | Polyclonal | 
| Conjugate | Unconjugated | 
| Host Species | Rabbit | 
Description
ZNF791 Polyclonal specifically detects ZNF791 in Human samples. It is validated for Western Blot.Specifications
| ZNF791 | |
| Polyclonal | |
| Rabbit | |
| NP_699189 | |
| 163049 | |
| Synthetic peptide directed towards the middle region of human ZNF791. Peptide sequence: CKECGKAFSLHSSFQRHTRIHNYEKPLECKQCGKAFSVSTSLKKHMRMHN | |
| Primary | 
| Western Blot | |
| Unconjugated | |
| RUO | |
| FLJ90396, MGC126179, MGC126180, zinc finger protein 791 | |
| ZNF791 | |
| IgG | 
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title