Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF799 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541252
Description
Peptide sequence: VIMGHSSLNC YIRVGAGHKP YEYHECGEKP DTHKQRGKAF SYHNSLQTHE Sequence homology: Human: 100%.
ZNF799 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 18 C2H2-type zinc fingers and 1 KRAB domain. ZNF799 may be involved in transcriptional regulation.
Specifications
ZNF799 | |
Polyclonal | |
Unconjugated | |
ZNF799 | |
HIT-40; zinc finger protein 14; zinc finger protein 799; Zinc finger protein 842; zinc finger protein HIT-40; ZNF799; ZNF842 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
90576 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96GE5 | |
ZNF799 | |
Synthetic peptide directed towards the N-terminal of human ZNF799 (aa 111-160). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction