Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF8 Monoclonal Antibody (2C6)
GREENER_CHOICE

Catalog No. PIMA524424
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524424 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524424 Supplier Invitrogen™ Supplier No. MA524424
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: ELWVAERGTT QGCHPAWEPR SESQASRKEE GLPEEEPSHV TGREGFPTDA PYPTTLGKDR ECQSQSLALK EQNNLKQLEF GLKEAPVQDQ GYKTLRLRE.

Transcriptional repressor. May modulate BMP and TGF-beta signal transduction, through its interaction with SMAD proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF8
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2C6
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ZNF8
Gene Accession No. P17098
Gene Alias HF.18; Zfp128; zinc finger protein 272; zinc finger protein 8; zinc finger protein 8 (clone HF.18); zinc finger protein HF.18; ZNF8
Gene Symbols ZNF8
Host Species Mouse
Immunogen ZNF8 (NP_066575.1, 82 a.a. ~ 180 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7554
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.