Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557061
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557061 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557061 Supplier Invitrogen™ Supplier No. PA557061
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LKEAPVQDQG YKTLRLRENC VLSSSPNPFP EISRGEYLYT YDSQITDSEH NSSLVSQQTG SPGKQPGENS DCHRDSSQAI PITELTK Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 55%.

Transcriptional repressor. May modulate BMP and TGF-beta signal transduction, through its interaction with SMAD proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF8
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF8
Gene Accession No. P17098
Gene Alias HF.18; Zfp128; zinc finger protein 272; zinc finger protein 8; zinc finger protein 8 (clone HF.18); zinc finger protein HF.18; ZNF8
Gene Symbols ZNF8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF8. Recombinant protein control fragment (Product #RP-97590).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7554
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.