Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF800 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554693
Description
Immunogen sequence: PIETNQNAVF QYISRTDNPI EVTESSSTPE QTEVQIQETS TEQSKTVPVT DTEVETVEPP PVEIVTDEVA PTSDEQPQES QADLETSDNS DFGHQLICCL CRKEFNSRRG VRRHIRKVHK E Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 84%.
May be involved in transcriptional regulation.
Specifications
ZNF800 | |
Polyclonal | |
Unconjugated | |
ZNF800 | |
PP902; Zinc finger protein 800; ZNF800 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
168850 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q2TB10 | |
ZNF800 | |
Recombinant protein corresponding to Human ZNF800. Recombinant protein control fragment (Product #RP-92617). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction