Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF800 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565708
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565708 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565708 Supplier Invitrogen™ Supplier No. PA565708
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IRHITVVHKKS SRYLGKITAS LEIRAIKKPI DFVLNKVAKR GPSRDEAKHS DSKHDGTSNS PSKKYEVADV GIEVKVTKNF SLHRCNKCGK AFAKKTYLEH HKKTHKANAS NSPEGNKTK Highest antigen sequence identity to the following orthologs - mouse 97%, rat 98%.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF800
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF800
Gene Accession No. Q2TB10
Gene Alias PP902; Zinc finger protein 800; ZNF800
Gene Symbols ZNF800
Host Species Rabbit
Immunogen Recombinant Human ZNF800. Recombinant protein control fragment (Product #RP-104939).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 168850
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.