Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF81 Antibody (PCRP-ZNF81-2G2), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314088MFV500
Description
ZNF81 Monoclonal antibody specifically detects ZNF81 in Human samples. It is validated for Flow CytometrySpecifications
ZNF81 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
347344 | |
Store at 4C in the dark. | |
IgG2a |
Flow Cytometry | |
PCRP-ZNF81-2G2 | |
Flow Cytometry | |
dJ54B20.6, FLJ44367, HFZ20X-linked 45, zinc finger protein 81, zinc finger protein 81 (HFZ20) | |
Recombinant full-length human ZNF81 protein (Uniprot: P51508) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction