Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF81 Mouse anti-Human, Clone: PCRP-ZNF81-2C7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP31394420UG
This item is not returnable.
View return policy
Description
ZNF81 Monoclonal antibody specifically detects ZNF81 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Protein ArraySpecifications
ZNF81 | |
Monoclonal | |
0.2 mg/ml | |
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array | |
P51508 | |
Mouse | |
Protein A or G purified | |
RUO | |
347344 | |
Store at 4°C. | |
IgG2b |
Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Peptide Array | |
PCRP-ZNF81-2C7 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
dJ54B20.6, FLJ44367, HFZ20X-linked 45, zinc finger protein 81, zinc finger protein 81 (HFZ20) | |
Recombinant full-length human ZNF81 protein (Uniprot: P51508) | |
20 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction