Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF81 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551625
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551625 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551625 Supplier Invitrogen™ Supplier No. PA551625
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VHPSPNLILS QKRPHKRDSF GKSFKHNLDL HIHNKSNAAK NLDKTIGHGQ VFTQNSSYSH HENTHTGVKF CERNQCGKVL SLKHSLSQNV KFPIGEKANT CT Highest antigen sequence identity to the following orthologs: Mouse - 34%, Rat - 50%.

This gene encodes a protein that likely functions as a transcription factor. The protein contains an N-terminal KRAB domain and several C2H2-type zinc finger motifs. Mutations in this gene cause mental retardation X-linked type 45 (MRX45).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF81
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF81
Gene Accession No. P51508
Gene Alias dJ54B20.6; HFZ20; MRX45; zinc finger protein 41-like; zinc finger protein 81; ZNF81
Gene Symbols ZNF81
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF81. Recombinant protein control fragment (Product #RP-91998).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 347344
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.