Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF816 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559986
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559986 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559986 Supplier Invitrogen™ Supplier No. PA559986
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RNLEFVDSSL KSMMEFSSTR HSITGEVI Highest antigen sequence identity to the following orthologs: Mouse - 36%, Rat - 36%.

ZNF816 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF816
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF816
Gene Accession No. Q0VGE8
Gene Alias Zinc finger protein 816; zinc finger protein 816A; ZNF816; ZNF816A
Gene Symbols ZNF816
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF816. Recombinant protein control fragment (Product #RP-98929).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 125893
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.