Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF821 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557631
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557631 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557631 Supplier Invitrogen™ Supplier No. PA557631
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TNPNKVHCDS EGDEEETTQD EVSSHTSEED GGVVKVEKEL ENTEQPVGGN EVVEHEVTGN LNSDPLLELC QCPLCQ Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 77%.

The ZNF821 protein contains two C2H2 zinc finger motifs and a score-and-three (23)-amino acid peptide repeat (STPR) domain. The STPR domain of the encoded protein binds to double stranded DNA and may also contain a nuclear localization signal, suggesting that this protein interacts with chromosomal DNA. The exact function of ZNF821, however, remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF821
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF821
Gene Accession No. O75541
Gene Alias 4930566A11Rik; Zfp821; Zinc finger protein 821; Znf821
Gene Symbols ZNF821
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF821. Recombinant protein control fragment (Product #RP-95503).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55565
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.