Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF823 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299212AF405
Description
ZNF823 Polyclonal specifically detects ZNF823 in Human samples. It is validated for Western Blot.Specifications
ZNF823 | |
Polyclonal | |
Western Blot | |
HSZFP36, ZFP 36 for a zinc finger protein, ZFP36, ZFP-36 for a zinc finger protein, zinc finger protein 823, zinc finger protein ZFP-36 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human ZNF823. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
55552 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction