Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF823 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB326513 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB326513 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB326513 Supplier Novus Biologicals Supplier No. NBP299212F
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF823 Polyclonal antibody specifically detects ZNF823 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen ZNF823
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate FITC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias HSZFP36, ZFP 36 for a zinc finger protein, ZFP36, ZFP-36 for a zinc finger protein, zinc finger protein 823, zinc finger protein ZFP-36
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human ZNF823.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55552
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.