Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF823 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543572
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543572 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543572 Supplier Invitrogen™ Supplier No. PA543572
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: GECGEVVLGH SSLNCNIRVD TGHKSCEHQE YGEKPYTHKQ RGKAISHQHS Sequence homology: Human: 100%.

cDNA sequence of HSZFP36 is generated by Mammalian Gene Collection (MGC) Program Team.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF823
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF823
Gene Accession No. P16415
Gene Alias HSZFP36; ZFP 36 for a zinc finger protein; ZFP36; Zinc finger protein 823; zinc finger protein ZFP-36; ZNF823
Gene Symbols ZNF823
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human HSZFP36 (aa 101-150).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55552
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.