Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF823 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540618
Description
Peptide sequence: GKAFSLAGSL RRHEATHTGV KPYKCQCGKA FSDLSSFQNH ETTHTGEKPY Sequence homology: Human: 100%; Rat: 79%.
cDNA sequence of HSZFP36 is generated by Mammalian Gene Collection (MGC) Program Team.
Specifications
ZNF823 | |
Polyclonal | |
Unconjugated | |
ZNF823 | |
HSZFP36; ZFP 36 for a zinc finger protein; ZFP36; Zinc finger protein 823; zinc finger protein ZFP-36; ZNF823 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55552 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P16415 | |
ZNF823 | |
Synthetic peptide directed towards the middle region of human HSZFP36. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction