Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF828 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989194
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9194 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9194 Supplier Bioss Supplier No. BS9607R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Required for proper alignment of chromosomes at metaphase and their accurate segregation during mitosis. Involved in the maintenance of spindle microtubules attachment to the kinetochore during sister chromatid biorientation. May recruit CENPE and CENPF to the kinetochore.

Specifications

Antigen ZNF828
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CHAMP1
Gene Accession No. Q96JM3
Gene Alias AA675043; AI116001; C13orf8; CAMP; CHAMP; Champ1; chromosome alignment maintaining phosphoprotein 1; Chromosome alignment-maintaining phosphoprotein 1; D8Ertd457e; D8Ertd569e; Kiaa1802; mKIAA1802; MRD40; Zfp828; Zinc finger protein 828; ZNF828
Gene Symbols CHAMP1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ZNF828/C13orf8.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283489
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.