Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF83 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555281
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555281 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555281 Supplier Invitrogen™ Supplier No. PA555281
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HGRKDDAQKQ PVKNQLGLNP QSHLPELQLF QAEGKIYKYD HMEKSVNSSS LVSPPQRISS TVKTHISHTY ECNFVDSLFT QKEKANIGTE HYKCSERGKA FHQGLHFTIH QIIHTKETQF KCDICGKIFN KKSNLASHQR IHTGEKPYKC Highest antigen sequence identity to the following orthologs: Mouse - 40%, Rat - 37%.

The ZNF83 gene encodes a zinc-finger protein belonging to a large family of transcription factors that play essential roles in regulating gene expression by interacting with DNA. Located on chromosome 3, ZNF83 is particularly notable for its recurrent mutations in cancer. For instance, the p.E293V missense mutation in ZNF83 has been frequently observed in bladder cancer patients, accounting for 18% of cases, and is associated with poor prognosis. This mutation disrupts the gene's tumor suppressor function, promoting increased tumor growth. Due to its significant role in tumorigenesis, ZNF83 is actively studied in cancer research as a potential biomarker and therapeutic target.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF83
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF83
Gene Accession No. P51522
Gene Alias HPF1; zinc finger protein 816B; Zinc finger protein 83; zinc finger protein HPF1; ZNF816B; ZNF83
Gene Symbols ZNF83
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF83. Recombinant protein control fragment (Product #RP-89397).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55769
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.