Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF830 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555170
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555170 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555170 Supplier Invitrogen™ Supplier No. PA555170
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QGKEHSVSSS REVTSSVLPN DFFSTNPPKA PIIPHSGSIE KAEIHEKVVE RRENTAEALP EGFFDDPEVD ARVRKVDAPK DQM Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 87%.

Located on chromosome 17, this gene encodes for a coiled-coil domain containing 16 protein with unknown function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF830
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF830
Gene Accession No. Q3MHS2, Q8R1N0, Q96NB3
Gene Alias 2410003C20Rik; AV301118; AW048207; CCDC16; coiled-coil domain containing 16; coiled-coil domain-containing protein 16; hypothetical protein MGC20398; MGC20398; Omcg1; orphan maintenance of genome 1; Ovus mutant candidate gene 1 protein; Zfp830; Zinc finger protein 830; Zinc finger protein 830-like protein; Znf830
Gene Symbols Zfp830, ZNF830
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF830. Recombinant protein control fragment (Product #RP-93551).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 497967, 66983, 91603
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.