Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF830 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541197
Description
Peptide sequence: ENTAEALPEG FFDDPEVDAR VRKVDAPKDQ MDKEWDEFQK AMRQVNTISE Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rat: 93%.
Located on chromosome 17, this gene encodes for a coiled-coil domain containing 16 protein with unknown function.
Specifications
| ZNF830 | |
| Polyclonal | |
| Unconjugated | |
| ZNF830 | |
| 2410003C20Rik; AV301118; AW048207; CCDC16; coiled-coil domain containing 16; coiled-coil domain-containing protein 16; hypothetical protein MGC20398; MGC20398; Omcg1; orphan maintenance of genome 1; Ovus mutant candidate gene 1 protein; Zfp830; Zinc finger protein 830; Zinc finger protein 830-like protein; Znf830 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 91603 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96NB3 | |
| ZNF830 | |
| Synthetic peptide directed towards the middle region of human CCDC16. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction