Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF835 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562327
Description
Immunogen sequence: LQGAELEGNW KHEGQVEDLQ ENQESCPEPE AVACKGDPAG DSMQERDEFS RIPRTISSPA ATQASVPDDS SSRR Highest antigen sequence identity to the following orthologs: Mouse - 26%, Rat - 26%.
May be involved in transcriptional regulation.
Specifications
ZNF835 | |
Polyclonal | |
Unconjugated | |
ZNF835 | |
BC37295_3; zinc finger protein 835; ZNF835 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
90485 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y2P0 | |
ZNF835 | |
Recombinant protein corresponding to Human ZNF835. Recombinant protein control fragment (Product #RP-101249). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction