Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF837 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$204.00 - $482.50
Specifications
Antigen | ZNF837 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP19140220
|
Novus Biologicals
NBP19140220UL |
20 μL |
Each for $204.00
|
|
|||||
NBP191402
|
Novus Biologicals
NBP191402 |
100 μL |
Each for $482.50
|
|
|||||
Description
ZNF837 Polyclonal specifically detects ZNF837 in Human samples. It is validated for Western Blot.Specifications
ZNF837 | |
Polyclonal | |
Rabbit | |
Human | |
zinc finger protein 837 | |
ZNF837 | |
IgG | |
58 kDa |
Western Blot | |
Unconjugated | |
RUO | |
XP_375665 | |
116412 | |
Synthetic peptide directed towards the N terminal of human LOC116412. Peptide sequence MEAPAQKAGQGGLPKADAQGASGAREKRPEEPRPLEEDRAGSRPTQKGDL. | |
Primary |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title