Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF84 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555992
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555992 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555992 Supplier Invitrogen™ Supplier No. PA555992
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLGYEVMKPD VIFKLEQGEE PWVGDGEIPS SDSPEVWKVD GNMMWHQDNQ DKLKIIKRGH ECDAFGKNFN LNMNFVPLRK SNSEGDLDGL ILKHHLDLLI PKGDYGKAES DDFNVFDNFF LHSKPEDTDT WLKYYDCDKY KES Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 28%.

ZNF84, a gene located on chromosome 12, encodes a zinc finger protein whose function is undescribed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF84
Applications ChIP Assay, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF84
Gene Accession No. P51523
Gene Alias HPF2; zinc finger protein 84; Zinc finger protein HPF2; ZNF84
Gene Symbols ZNF84
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF84. Recombinant protein control fragment (Product #RP-99736).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7637
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.