Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF84 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541095
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541095 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541095 Supplier Invitrogen™ Supplier No. PA541095
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RKAFSQKSQL VNHQRIHTGE KPYRCIECGK AFSQKSQLIN HQRTHTVKKS Sequence homology: Cow: 100%; Dog: 100%; Horse: 93%; Human: 100%; Mouse: 92%; Pig: 90%; Rabbit: 100%; Rat: 83%.

ZNF84, a gene located on chromosome 12, encodes a zinc finger protein whose function is undescribed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF84
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF84
Gene Accession No. P51523
Gene Alias HPF2; zinc finger protein 84; Zinc finger protein HPF2; ZNF84
Gene Symbols ZNF84
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ZNF84 (aa 689-738).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7637
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.