Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF841 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA548212

Catalog No. PA548212


Only null left
Add to Cart

Description

Description

ZNF841 Polyclonal Antibody for Western Blot, Flow

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZNF841
Polyclonal
PBS with 0.09% sodium azide; pH 7.4
Q6ZN19
ZNF841
KLH conjugated synthetic peptide between 34-63 amino acids from the N-terminal region of human ZNF841.
400 μL
Primary
Human
Antibody
IgG
Flow Cytometry, Western Blot
Unconjugated
ZNF841
Zinc finger protein 841; ZNF841
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
284371
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.