Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF846 Antibody (PCRP-ZNF846-1E12), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP32047120UG
Description
ZNF846 Monoclonal antibody specifically detects ZNF846 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
ZNF846 | |
Monoclonal | |
0.2 mg/ml | |
Western Blot 1-2 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL | |
zinc finger protein 846 | |
Recombinant full-length human ZNF846 protein | |
20 μg | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin) | |
PCRP-ZNF846-1E12 | |
Unconjugated | |
10mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
162993 | |
Store at 4&drg;C. Do not freeze. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction