Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF846 Antibody (ZNF846/2687), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308846AF488
Description
ZNF846 Monoclonal antibody specifically detects ZNF846 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein ArraySpecifications
ZNF846 | |
Monoclonal | |
Alexa Fluor 488 | |
zinc finger protein 846 | |
Recombinant full-length human ZNF846 protein (Uniprot: Q147U1) | |
100 μg | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin), Protein Array | |
ZNF846/2687 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
162993 | |
Store at 4°C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction