Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF852 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561709
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561709 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561709 Supplier Invitrogen™ Supplier No. PA561709
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YKEVEEHPPL SSSPVEHEGV LKGQKSYRCD E Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 52%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF852
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF852
Gene Accession No. Q6ZMS4
Gene Alias putative zinc finger protein 852; zinc finger protein 852; ZNF852
Gene Symbols ZNF852
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF852. Recombinant protein control fragment (Product #RP-99541).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285346
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.