Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566556
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566556 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566556 Supplier Invitrogen™ Supplier No. PA566556
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GMRSRGRGGFT SDRGFQFVSS SLPDICYRCG ESG Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

This gene encodes a nucleic-acid binding protein with seven zinc-finger domains. The protein has a preference for binding single stranded DNA and RNA. The protein functions in cap-independent translation of ornithine decarboxylase mRNA, and may also function in sterol-mediated transcriptional regulation. A CCTG expansion in the first intron of this gene results in myotonic dystrophy type 2. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF9
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CNBP
Gene Accession No. P62633
Gene Alias AA408710; CCHC-type zinc finger nucleic acid binding protein; CCHC-type zinc finger, nucleic acid binding protein; cellular nucleic acid binding protein; cellular nucleic acid binding protein 1; cellular nucleic acid-binding protein; CNBP; CNBP1; DM2; erythroid differentiation-related; HGNC:13164; PROMM; RNF163; sterol regulatory element-binding protein; ZCCHC22; zinc finger protein 273; zinc finger protein 9; zinc finger protein 9 (a cellular retroviral nucleic acid binding protein); Znf9
Gene Symbols CNBP
Host Species Rabbit
Immunogen Recombinant Human ZNF9. Recombinant protein control fragment (Product #RP-105254).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7555
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.