Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590229
Description
Positive Samples: HeLa, A-549, LO2, mouse brain, mouse kidney, mouse liver; Cellular Location: Cytoplasm, Endoplasmic reticulum Immunogen sequence: CGRGGHIAKD CKEPKREREQ CCYNCGKPGH LARDCDHADE QKCYSCGEFG HIQKDCTKVK CYRCGETGHV AINCSKTSEV NCYRCGESGH LARECTIEAT A.
This gene encodes a nucleic-acid binding protein with seven zinc-finger domains. The protein has a preference for binding single stranded DNA and RNA. The protein functions in cap-independent translation of ornithine decarboxylase mRNA, and may also function in sterol-mediated transcriptional regulation. A CCTG expansion in the first intron of this gene results in myotonic dystrophy type 2. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
ZNF9 | |
Polyclonal | |
Unconjugated | |
CNBP | |
AA408710; CCHC-type zinc finger nucleic acid binding protein; CCHC-type zinc finger, nucleic acid binding protein; cellular nucleic acid binding protein; cellular nucleic acid binding protein 1; cellular nucleic acid-binding protein; CNBP; CNBP1; DM2; erythroid differentiation-related; HGNC:13164; PROMM; RNF163; sterol regulatory element-binding protein; ZCCHC22; zinc finger protein 273; zinc finger protein 9; zinc finger protein 9 (a cellular retroviral nucleic acid binding protein); Znf9 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
12785, 7555 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P53996, P62633 | |
CNBP | |
A synthetic peptide corresponding to a sequence within amino acids 70-170 of human CNBP (NP_0011206681). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction