Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF9 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518505
Description
This antibody is predicted to react with canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
This gene encodes a nucleic-acid binding protein with seven zinc-finger domains. The protein has a preference for binding single stranded DNA and RNA. The protein functions in cap-independent translation of ornithine decarboxylase mRNA, and may also function in sterol-mediated transcriptional regulation. A CCTG expansion in the first intron of this gene results in myotonic dystrophy type 2. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| ZNF9 | |
| Polyclonal | |
| Unconjugated | |
| CNBP | |
| AA408710; CCHC-type zinc finger nucleic acid binding protein; CCHC-type zinc finger, nucleic acid binding protein; cellular nucleic acid binding protein; cellular nucleic acid binding protein 1; cellular nucleic acid-binding protein; CNBP; CNBP1; DM2; erythroid differentiation-related; HGNC:13164; PROMM; RNF163; sterol regulatory element-binding protein; ZCCHC22; zinc finger protein 273; zinc finger protein 9; zinc finger protein 9 (a cellular retroviral nucleic acid binding protein); Znf9 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 12785, 7555 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P53996, P62633 | |
| CNBP | |
| Synthetic peptide sequence (GESGHLARECTIE) corresponding to the C-terminus amino acids of CNBP (aa 162-175). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction