Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF90 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5107324

Catalog No. PIPA5107324


Only null left
Add to Cart

Description

Description

ZNF90 Polyclonal Antibody for Western Blot

ZNF90 may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZNF90
Polyclonal
Unconjugated
ZNF90
HTF9; Zinc finger protein 90; zinc finger protein HTF9; ZNF90
Rabbit
Affinity chromatography
RUO
7643
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Q03938
ZNF90
Synthetic peptide directed towards the N terminal region of human ZNF90.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.