Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF91 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $499.50
Specifications
| Antigen | ZNF91 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17922620
![]() |
Novus Biologicals
NBP17922620UL |
20 μL |
Each for $206.00
|
|
|||||
NBP179226
![]() |
Novus Biologicals
NBP179226 |
100 μL |
Each for $499.50
|
|
|||||
Description
ZNF91 Polyclonal specifically detects ZNF91 in Human samples. It is validated for Western Blot.Specifications
| ZNF91 | |
| Polyclonal | |
| Rabbit | |
| NP_003421 | |
| 7644 | |
| Synthetic peptide directed towards the N terminal of human ZNF91The immunogen for this antibody is ZNF91. Peptide sequence SNSSTLANHKITHTEEKPYKCKECDKAFKRLSTLTKHKIIHAGEKLYKCE. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| HPF7, HTF10, zinc finger protein 91, zinc finger protein 91 (HPF7, HTF10), zinc finger protein HPF7, zinc finger protein HTF10 | |
| ZNF91 | |
| IgG | |
| 137 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title