Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF91 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543064
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543064 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543064 Supplier Invitrogen™ Supplier No. PA543064
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: HKIIHAGEKL YKCEECGKAF NRSSNLTIHK FIHTGEKPNW Sequence homology: Human: 100%; Mouse: 93%; Rat: 93%.

ZNF91 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 36 C2H2-type zinc fingers and 1 KRAB domain. The function of the ZNF91 protein remains unknown. Western blots using two different antibodies against two unique regions of this protein target confirm the same apparent molecular weight in our tests.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF91
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF91
Gene Accession No. Q05481
Gene Alias HPF7; HTF10; Zinc finger protein 91; zinc finger protein 91 (HPF7, HTF10); zinc finger protein HPF7; Zinc finger protein HTF10; ZNF91
Gene Symbols ZNF91
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ZNF91.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7644
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.