Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF93 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116330
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116330 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116330 Supplier Invitrogen™ Supplier No. PA5116330
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF93 Polyclonal Antibody for Western Blot

ZNF93 belongs to the krueppel C2H2-type zinc-finger protein family and may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF93
Applications Western Blot
Classification Polyclonal
Concentration 0.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ZNF93
Gene Accession No. P35789
Gene Alias HPF34; HTF34; TF34; Zinc finger protein 505; zinc finger protein 93; Zinc finger protein HTF34; ZNF354B; ZNF505; ZNF93
Gene Symbols ZNF93
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 40-67 amino acids from the N-terminal region of human ZNF93.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81931
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.