Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF93 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982974
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2974 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2974 Supplier Bioss Supplier No. BS13558R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF93 belongs to the krueppel C2H2-type zinc-finger protein family and may be involved in transcriptional regulation.

Specifications

Antigen ZNF93
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ZNF93
Gene Accession No. P35789
Gene Alias BOS_7081; HPF34; HTF34; LOC510078; TF34; zinc finger protein 354A-like; Zinc finger protein 505; Zinc finger protein 93; Zinc finger protein HTF34; ZNF505; ZNF93
Gene Symbols ZNF93
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ZBT24/ZNF45.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81931
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.