Learn More
Invitrogen™ ZNF96 Monoclonal Antibody (3E4)

Description
Peptide Sequence: MASTWAIQAH MDQDEPLEVK IEEEKYTTRQ DWDLRKNNTH SREVFRQYFR QFCYQETSGP REALSRLREL CHQWLRPETH TKEQILELLV LEQFLTILPE ELQAWVQEQH PESGEEVVTV LEDLERELDE PGEQVSVHTG EQEMFLQETV RLRKEGEPSM SLQSMKAQPK YESPELESQQ EQVLDVETGN EYGNLKQEVS EEMEPHGKTS SKFENDMSKS ARCGETREPE EITEEPSACS REDKQPTCDE NGVSLTENSD HTEHQRICPG EESYGCDDCG KAFSQHSHLI EHQRIHTGDR PYKCEECGKA FRGRTVLIRH KIIHTGEKPY KCNECGKAFG RWSALNQHQR LHTGEKHYHC NDCGKAFSQK AGLFHHIKIH TRDKPYQCTQ CNKSFSRRSI LTQHQGVHTG AKPYECNECG KAFVYNSSLV SHQEIHHKEK CYQCKECGKS FSQSGLIQHQ RIHTGEKPYK CDVCEKAFIQ RTSLTEHQRI HTGERPYKCD KCGKAFTQRS VLTEHQRIEC GNAFRGITSL IQHQRIHTGE KPYQCDECGK AFRQRKKTSY KEILLKNHSE PQAGVNLLLS SLIPEWQSCF RRDL.
Specifications
Specifications
| Antigen | ZNF96 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 3E4 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ZSCAN12 |
| Gene Accession No. | O43309 |
| Gene Alias | 2510038J07Rik; dJ29K1.2; DM9; FPM315; LOW QUALITY PROTEIN: zinc finger and SCAN domain-containing protein 12; mKIAA0426; Zfp96; Zfp-96; zinc finger and SCAN domain containing 12; zinc finger and SCAN domain containing 23; zinc finger and SCAN domain-containing protein 12; zinc finger protein 305; zinc finger protein 323; zinc finger protein 96; ZNF29K1; ZNF305; ZNF323; ZNF96; Zscan12; ZSCAN23 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.