Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF96 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568827
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568827 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568827 Supplier Thermo Scientific Supplier No. PA568827
Add to Cart
Add to Cart

ZNF96 Polyclonal Antibody

ZNF96 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF96
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZSCAN12
Gene Accession No. Q9Z1D7
Gene Alias 2510038J07Rik; dJ29K1.2; DM9; FPM315; LOW QUALITY PROTEIN: zinc finger and SCAN domain-containing protein 12; mKIAA0426; Zfp96; Zfp-96; zinc finger and SCAN domain containing 12; zinc finger and SCAN domain containing 23; zinc finger and SCAN domain-containing protein 12; zinc finger protein 305; zinc finger protein 323; zinc finger protein 96; ZNF29K1; ZNF305; ZNF323; ZNF96; Zscan12; ZSCAN23
Gene Symbols ZSCAN12
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse ZNF96 (aa 421-470).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22758
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.