Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNHIT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599756
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599756 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599756 Supplier Invitrogen™ Supplier No. PA599756
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ZNHIT1.

Seems to play a role in p53-mediated apoptosis induction (PubMed:17380123). Binds to NR1D2 and relieves it of its inhibitory effect on the transcription of APOC3 without affecting its DNA-binding activity (PubMed:17892483). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNHIT1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ZNHIT1
Gene Accession No. O43257, Q8R331
Gene Alias 2700001K05Rik; CG1I; CGBP1; Cyclin-G1-binding protein 1; H_DJ0747G18.14; hypothetical protein LOC407699; p18 Hamlet; p18Hamlet; putative cyclin G1 interacting protein; similar to Zinc finger HIT domain containing protein 1 (Cyclin G1-binding protein 1); zgc:112524; zgc:112524 protein; zinc finger HIT domain-containing protein 1; zinc finger HIT-type containing 1; zinc finger protein subfamily 4A member 1; zinc finger protein, subfamily 4A (HIT domain containing), member 1; zinc finger, HIT domain containing 1; zinc finger, HIT type 1; zinc finger, HIT-type containing 1; ZNFN4A1; ZNHIT1
Gene Symbols ZNHIT1
Host Species Rabbit
Immunogen A synthesized peptide derived from human ZNHIT1(Accession O43257), corresponding to amino acid residues R21-T71.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100360077, 10467, 70103
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.