Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNHIT2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP215179
Description
ZNHIT2 Polyclonal specifically detects ZNHIT2 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
ZNHIT2 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
C11orf5zinc finger HIT domain-containing protein 2, chromosome 11 open reading frame 5, FONMGC120286, MGC120285, Protein FON, zinc finger, HIT domain containing 2, zinc finger, HIT type 2, zinc finger, HIT-type containing 2 | |
Rabbit | |
43 kDa | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
1mg/mL | |
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
Q9UHR6 | |
ZNHIT2 | |
Recombinant protein encompassing a sequence within the center region of human ZNHIT2. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
741 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction