Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNHIT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563517
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563517 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563517 Supplier Invitrogen™ Supplier No. PA563517
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CTPVVPTRIP AIVSLSRGPV SPLVRFQLPN VLFAYAHTLA LYHGGDDALL SDFCATLLGV SGALGAQQVF ASAEEAL Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 82%.

The ZNHIT2 gene, located on chromosome 7, encodes a zinc-finger HIT-type containing protein crucial for various cellular processes, such as chromatin remodeling and gene transcription regulation. Knockout studies in mice reveal that ZNHIT2, alongside ZNHIT1, is essential during early development, with deficits leading to embryonic lethality during peri-implantation stages due to reduced proportions of pluripotent inner cell mass cells and aberrant expression of key developmental genes like NANOG and SOX17. This gene's relevance extends to its involvement in protein-protein and protein-nucleic acid interactions that are fundamental in cellular functions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNHIT2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Znhit2
Gene Accession No. Q9UHR6
Gene Alias C11orf5; FON; ORF6; Protein FON; RGD1307918; zinc finger HIT domain-containing protein 2; zinc finger HIT-type containing 2; zinc finger, HIT domain containing 2; zinc finger, HIT domain containing 2, pseudogene; zinc finger, HIT type 2; zinc finger, HIT-type containing 2; Znhit2; Znhit2-ps
Gene Symbols Znhit2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNHIT2. Recombinant protein control fragment (Product #RP-99907).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 741
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.