Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNHIT3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566301
Description
Immunogen sequence: SDEEEDRVSLQ NLKNLGESAT LRSLLLNPHL RQLMVNLDQG EDKAKLMRAY MQEPLFVEFA DCCLGIVEPS Q Highest antigen sequence identity to the following orthologs - mouse 92%, rat 83%.
ZNHIT3 contains 1 HIT-type zinc finger. The exact function of ZNHIT3 remains unknown.
Specifications
ZNHIT3 | |
Polyclonal | |
Unconjugated | |
ZNHIT3 | |
HNF-4a coactivator; Thyroid hormone receptor interactor 3; thyroid receptor interacting protein 3; thyroid receptor-interacting protein 3; TR-interacting protein 3; TRIP3; TRIP-3; Zinc finger HIT domain-containing protein 3; zinc finger HIT domain-containing protein-like protein; zinc finger HIT type 3; zinc finger HIT-type containing 3; zinc finger, HIT domain containing 3; zinc finger, HIT type 3; zinc finger, HIT-type containing 3; ZNHIT3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9326 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15649 | |
ZNHIT3 | |
Recombinant Human ZNHIT3. Recombinant protein control fragment (Product #RP-106981). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction