Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNHIT3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543577
Description
Peptide sequence: HKEQCNPETR PVEKKIRSAL PTKTVKPVEN KDDDDSIADF LNSDEEEDRV Sequence homology: Dog: 93%; Horse: 93%; Human: 100%; Pig: 100%; Rabbit: 93%.
ZNHIT3 contains 1 HIT-type zinc finger. The exact function of ZNHIT3 remains unknown.
Specifications
ZNHIT3 | |
Polyclonal | |
Unconjugated | |
ZNHIT3 | |
HNF-4a coactivator; Thyroid hormone receptor interactor 3; thyroid receptor interacting protein 3; thyroid receptor-interacting protein 3; TR-interacting protein 3; TRIP3; TRIP-3; Zinc finger HIT domain-containing protein 3; zinc finger HIT domain-containing protein-like protein; zinc finger HIT type 3; zinc finger HIT-type containing 3; zinc finger, HIT domain containing 3; zinc finger, HIT type 3; zinc finger, HIT-type containing 3; ZNHIT3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
9326 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q15649 | |
ZNHIT3 | |
Synthetic peptide directed towards the N-terminal of human ZNHIT3 (aa 38-87). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction