Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNHIT6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578513
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578513 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578513 Supplier Invitrogen™ Supplier No. PA578513
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: MCF-7, MDA-MB-231 Predicted Reactivity: Mouse (85%), Rat (81%), Pig (91%), Rhesus Monkey (98%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Required for box C/D snoRNAs accumulation involved in snoRNA processing, snoRNA transport to the nucleolus and ribosome biogenesis. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNHIT6
Applications Western Blot
Classification Polyclonal
Concentration 0.16 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene ZNHIT6
Gene Accession No. Q9NWK9
Gene Alias 2410019A14Rik; BCD1; box C/D snoRNA essential 1 homolog; box C/D snoRNA protein 1; box C/d snoRNA protein 1-like protein; C1orf181; C3H1ORF181; FLJ20729; NY-BR-75; RGD1308723; Serologically defined breast cancer antigen NY-BR-75; zinc finger HIT domain-containing protein 6; zinc finger HIT-type containing 6; zinc finger, HIT type 6; zinc finger, HIT-type containing 6; Znhit6
Gene Symbols ZNHIT6
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ZNHIT6. The exact sequence is proprietary.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54680
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.