Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNHIT6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114925
Description
Antibody detects endogenous levels of total CA181.
Required for box C/D snoRNAs accumulation involved in snoRNA processing, snoRNA transport to the nucleolus and ribosome biogenesis. [UniProt].
Specifications
ZNHIT6 | |
Polyclonal | |
Unconjugated | |
ZNHIT6 | |
2410019A14Rik; BCD1; box C/D snoRNA essential 1 homolog; box C/D snoRNA protein 1; box C/d snoRNA protein 1-like protein; C1orf181; C3H1ORF181; FLJ20729; NY-BR-75; RGD1308723; Serologically defined breast cancer antigen NY-BR-75; zinc finger HIT domain-containing protein 6; zinc finger HIT-type containing 6; zinc finger, HIT type 6; zinc finger, HIT-type containing 6; Znhit6 | |
Rabbit | |
Affinity chromatography | |
RUO | |
229937, 54680 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q3UFB2, Q9NWK9 | |
ZNHIT6 | |
A synthesized peptide derived from human ZNHIT6(Accession Q9NWK9), corresponding to amino acid residues Y305-S355. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction